Assay Details
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030954.1 | NP_001026125.1 | 13 - 14 | 2060 | 55 | |
XM_015283100.2 | XP_015138586.1 | 15 - 16 | 2232 | 55 | ||
XM_015283101.2 | XP_015138587.1 | 15 - 16 | 2229 | 55 | ||
GenBank mRNA | AJ719272.1 | - | 13 - 14 | 2060 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
