Assay Details
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030974.1 | NP_001026145.1 | 2 - 3 | 338 | 95 | |
GenBank mRNA | AJ851572.1 | - | 2 - 3 | 338 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
