Assay Details
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031347.1 | NP_001026518.1 | 2 - 3 | 166 | 62 | |
XM_015299933.2 | XP_015155419.1 | 2 - 3 | 166 | 62 | ||
XM_015299934.2 | XP_015155420.1 | 2 - 3 | 2005 | 62 | ||
XM_015299935.2 | XP_015155421.1 | 2 - 3 | 195 | 62 | ||
XM_015299936.2 | XP_015155422.1 | 2 - 3 | 199 | 62 | ||
XM_015299937.2 | XP_015155423.1 | 2 - 3 | 166 | 62 | ||
XM_015299938.2 | XP_015155424.1 | 2 - 3 | 168 | 62 | ||
GenBank mRNA | AJ719636.1 | - | 2 - 3 | 166 | 62 | |
DQ126684.1 | - | 1 - 2 | 105 | 62 | ||
KC905084.1 | - | 1 - 2 | 105 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
