Assay Details
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031347.1 | NP_001026518.1 | 5 - 6 | 495 | 70 | |
XM_015299934.2 | XP_015155420.1 | 5 - 6 | 2334 | 70 | ||
XM_015299936.2 | XP_015155422.1 | 5 - 6 | 528 | 70 | ||
XM_015299938.2 | XP_015155424.1 | 5 - 6 | 497 | 70 | ||
GenBank mRNA | AJ719636.1 | - | 5 - 6 | 495 | 70 | |
KC905084.1 | - | 4 - 5 | 434 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
