Assay Details
Gene Symbol: RBM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031347.1 | NP_001026518.1 | 6 | 1232 | 120 | |
XM_015299933.2 | XP_015155419.1 | 6 | 1234 | 119 | ||
XM_015299934.2 | XP_015155420.1 | 6 | 3070 | 119 | ||
XM_015299935.2 | XP_015155421.1 | 7 | 846 | 119 | ||
XM_015299936.2 | XP_015155422.1 | 7 | 847 | 119 | ||
GenBank mRNA | AJ719636.1 | - | 6 | 1232 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
