Assay Details
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031406.2 | NP_001026577.1 | 3 - 4 | 639 | 64 | |
XM_015294667.1 | XP_015150153.1 | 3 - 4 | 634 | 64 | ||
GenBank mRNA | AJ720999.1 | - | 3 - 4 | 639 | 64 | |
BU208883.1 | - | 2 - 3 | 288 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
