Assay Details
Gene Symbol: IL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037825.1 | NP_001032914.1 | 1 - 2 | 405 | 69 | |
XM_015293972.2 | XP_015149458.1 | 2 - 3 | 1132 | 69 | ||
GenBank mRNA | AM773755.1 | - | 1 - 2 | 102 | 69 | |
BX931122.2 | - | 1 - 2 | 412 | 69 | ||
DQ294769.1 | - | 1 - 2 | 405 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
