Assay Details
Gene Symbol: TNFRSF13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037828.1 | NP_001032917.1 | 1 - 2 | 136 | 80 | |
GenBank mRNA | AM085437.1 | - | 1 - 2 | 136 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
