Assay Details
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037832.1 | NP_001032921.1 | 438 | 111 | ||
GenBank mRNA | AM076721.1 | - | 438 | 111 | ||
CR406241.1 | - | 481 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
