Assay Details
Gene Symbol: FOXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039284.2 | NP_001034373.2 | 3 - 4 | 735 | 100 | |
XM_015278070.2 | XP_015133556.1 | 4 - 5 | 777 | 100 | ||
XM_015278071.2 | XP_015133557.1 | 5 - 6 | 1188 | 100 | ||
XM_015278072.2 | XP_015133558.1 | 5 - 6 | 5481 | 100 | ||
XM_015278073.2 | XP_015133559.1 | 4 - 5 | 874 | 100 | ||
GenBank mRNA | AJ721059.1 | - | 4 - 5 | 778 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
