Assay Details
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001311.2 | NP_001001311.2 | 1 - 2 | 547 | 97 | |
GenBank mRNA | AY574987.1 | - | 1 - 2 | 547 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
