Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007085.2 | NP_001007086.1 | 2 - 3 | 194 | 81 | |
GenBank mRNA | AJ621250.1 | - | 2 - 3 | 194 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
