Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 2 - 3 | 241 | 86 | |
GenBank mRNA | CD527028.1 | - | 2 - 3 | 155 | 86 | |
CR406506.1 | - | 2 - 3 | 250 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
