Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 4 - 5 | 445 | 83 | |
GenBank mRNA | BU204430.1 | - | 2 - 3 | 122 | 83 | |
CD527028.1 | - | 4 - 5 | 359 | 83 | ||
CR406506.1 | - | 4 - 5 | 454 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
