Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 5 - 6 | 641 | 114 | |
GenBank mRNA | BU204430.1 | - | 3 - 4 | 318 | 114 | |
BU326578.1 | - | 93 | 113 | |||
CD527028.1 | - | 5 - 6 | 555 | 114 | ||
CR406506.1 | - | 5 - 6 | 650 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
