Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 6 - 7 | 713 | 136 | |
GenBank mRNA | BU204430.1 | - | 4 - 5 | 390 | 136 | |
CR406506.1 | - | 6 - 7 | 722 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
