Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012653.2 | NP_001012671.1 | 7 - 8 | 880 | 85 | |
GenBank mRNA | BQ038887.2 | - | 1 - 2 | 45 | 85 | |
BU204430.1 | - | 5 - 6 | 557 | 85 | ||
BU425613.1 | - | 2 - 3 | 199 | 85 | ||
BX275259.3 | - | 2 - 3 | 201 | 85 | ||
CR406506.1 | - | 7 - 8 | 889 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
