Assay Details
Gene Symbol: OLFM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040463.1 | NP_001035553.1 | 5 - 6 | 675 | 86 | |
GenBank mRNA | AB236174.1 | - | 5 - 6 | 675 | 86 | |
CR390719.1 | - | 4 - 5 | 539 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
