Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044653.1 | NP_001038118.1 | 10 - 11 | 1630 | 81 | |
XM_015287980.1 | XP_015143466.1 | 11 - 12 | 1777 | 81 | ||
GenBank mRNA | AB255032.1 | - | 3 - 4 | 380 | 81 | |
AB295599.1 | - | 10 - 11 | 1368 | 81 | ||
AY882944.1 | - | 10 - 11 | 1630 | 81 | ||
EU124632.1 | - | 10 - 11 | 1388 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
