Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044653.1 | NP_001038118.1 | 3 - 4 | 580 | 74 | |
XM_015287980.1 | XP_015143466.1 | 4 - 5 | 727 | 74 | ||
GenBank mRNA | AB255031.1 | - | 3 - 4 | 318 | 74 | |
AB295599.1 | - | 3 - 4 | 318 | 74 | ||
AY882944.1 | - | 3 - 4 | 580 | 74 | ||
EU124632.1 | - | 3 - 4 | 338 | 74 | ||
EU124633.1 | - | 3 - 4 | 338 | 74 | ||
EU124634.1 | - | 3 - 4 | 338 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
