Assay Details
Gene Symbol: OPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044653.1 | NP_001038118.1 | 8 - 9 | 1402 | 132 | |
XM_015287980.1 | XP_015143466.1 | 9 - 10 | 1549 | 132 | ||
GenBank mRNA | AB255032.1 | - | 1 - 2 | 152 | 132 | |
AB295599.1 | - | 8 - 9 | 1140 | 132 | ||
AY882944.1 | - | 8 - 9 | 1402 | 132 | ||
EU124632.1 | - | 8 - 9 | 1160 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
