Assay Details
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044674.1 | NP_001038139.1 | 10 - 11 | 1174 | 144 | |
XM_015277359.1 | XP_015132845.1 | 10 - 11 | 1333 | 144 | ||
XM_015277360.2 | XP_015132846.1 | 10 - 11 | 1265 | 144 | ||
XM_015277361.2 | XP_015132847.1 | 10 - 11 | 1169 | 144 | ||
XM_015277362.2 | XP_015132848.1 | 10 - 11 | 1108 | 144 | ||
XM_015277363.2 | XP_015132849.1 | 10 - 11 | 1158 | 144 | ||
GenBank mRNA | AB161354.1 | - | 10 - 11 | 1174 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
