Assay Details
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044674.1 | NP_001038139.1 | 11 - 12 | 1278 | 59 | |
XM_015277359.1 | XP_015132845.1 | 11 - 12 | 1437 | 59 | ||
XM_015277360.2 | XP_015132846.1 | 11 - 12 | 1369 | 59 | ||
XM_015277361.2 | XP_015132847.1 | 11 - 12 | 1273 | 59 | ||
XM_015277362.2 | XP_015132848.1 | 11 - 12 | 1212 | 59 | ||
XM_015277363.2 | XP_015132849.1 | 11 - 12 | 1262 | 59 | ||
GenBank mRNA | AB161354.1 | - | 11 - 12 | 1278 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
