Assay Details
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044674.1 | NP_001038139.1 | 2 - 3 | 232 | 87 | |
XM_015277359.1 | XP_015132845.1 | 2 - 3 | 391 | 87 | ||
XM_015277360.2 | XP_015132846.1 | 2 - 3 | 323 | 87 | ||
XM_015277361.2 | XP_015132847.1 | 2 - 3 | 227 | 87 | ||
XM_015277362.2 | XP_015132848.1 | 2 - 3 | 166 | 87 | ||
XM_015277363.2 | XP_015132849.1 | 2 - 3 | 216 | 87 | ||
GenBank mRNA | AB161354.1 | - | 2 - 3 | 232 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
