Assay Details
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 10 - 11 | 1258 | 125 | |
XM_015281391.2 | XP_015136877.1 | 8 - 9 | 1209 | 125 | ||
XM_015281392.2 | XP_015136878.1 | 12 - 13 | 3530 | 125 | ||
GenBank mRNA | AB243875.1 | - | 10 - 11 | 1258 | 125 | |
AB243876.1 | - | 8 - 9 | 1071 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
