Assay Details
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 3 - 4 | 266 | 104 | |
XM_015281392.2 | XP_015136878.1 | 5 - 6 | 2538 | 104 | ||
GenBank mRNA | AB243875.1 | - | 3 - 4 | 266 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
