Assay Details
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048077.1 | NP_001041542.1 | 7 - 8 | 842 | 78 | |
XM_015281391.2 | XP_015136877.1 | 5 - 6 | 793 | 78 | ||
XM_015281392.2 | XP_015136878.1 | 9 - 10 | 3114 | 78 | ||
GenBank mRNA | AB243875.1 | - | 7 - 8 | 842 | 78 | |
AB243876.1 | - | 5 - 6 | 655 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
