Assay Details
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079479.2 | NP_001072947.1 | 3 - 4 | 306 | 67 | |
NM_001278150.1 | NP_001265079.1 | 3 - 4 | 306 | 67 | ||
XM_015277943.2 | XP_015133429.1 | 3 - 4 | 317 | 67 | ||
GenBank mRNA | AJ719476.1 | - | 3 - 4 | 306 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
