Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 10 - 11 | 1116 | 99 | |
GenBank mRNA | AB086192.2 | - | 10 - 11 | 1164 | 99 | |
AL584959.1 | - | 2 - 3 | 277 | 99 | ||
BX931986.1 | - | 10 - 11 | 1217 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
