Assay Details
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204400.2 | NP_989731.1 | 6 - 7 | 641 | 112 | |
GenBank mRNA | AB086192.2 | - | 6 - 7 | 689 | 112 | |
BX931986.1 | - | 6 - 7 | 742 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
