Assay Details
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113292.1 | NP_001106763.1 | 1 - 2 | 88 | 64 | |
XM_015287565.1 | XP_015143051.1 | 2 - 3 | 722 | 64 | ||
XM_025150841.1 | XP_025006609.1 | 3 - 4 | 887 | 64 | ||
XM_025150842.1 | XP_025006610.1 | 2 - 3 | 731 | 64 | ||
GenBank mRNA | EF621526.1 | - | 1 - 2 | 88 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
