Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204300.1 | NP_989631.1 | 14 - 15 | 2362 | 140 | |
XM_015283590.2 | XP_015139076.1 | 14 - 15 | 2477 | 140 | ||
XM_015283596.2 | XP_015139082.1 | 13 - 14 | 2416 | 140 | ||
XM_015283598.2 | XP_015139084.1 | 14 - 15 | 2618 | 140 | ||
XM_015283599.2 | XP_015139085.1 | 11 - 12 | 2041 | 140 | ||
XM_015283601.2 | XP_015139087.1 | 14 - 15 | 1782 | 140 | ||
XM_015283602.2 | XP_015139088.1 | 13 - 14 | 1713 | 140 | ||
XM_015283605.2 | XP_015139091.1 | 13 - 14 | 1856 | 140 | ||
XM_015283606.2 | XP_015139092.1 | 12 - 13 | 1598 | 140 | ||
XM_015283607.2 | XP_015139093.1 | 12 - 13 | 1742 | 140 | ||
XM_025148476.1 | XP_025004244.1 | 14 - 15 | 2431 | 140 | ||
XM_025148477.1 | XP_025004245.1 | 14 - 15 | 2422 | 140 | ||
XM_025148478.1 | XP_025004246.1 | 14 - 15 | 2395 | 140 | ||
XM_025148479.1 | XP_025004247.1 | 13 - 14 | 1661 | 140 | ||
GenBank mRNA | AB017437.1 | - | 13 - 14 | 1886 | 140 | |
AB035522.1 | - | 12 - 13 | 1775 | 140 | ||
AB035523.1 | - | 10 - 11 | 1480 | 140 | ||
AB057641.1 | - | 14 - 15 | 2362 | 140 | ||
CR386583.1 | - | 7 - 8 | 551 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
