Assay Details
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204300.1 | NP_989631.1 | 5 - 6 | 490 | 84 | |
XM_015283590.2 | XP_015139076.1 | 4 - 5 | 545 | 84 | ||
XM_015283596.2 | XP_015139082.1 | 4 - 5 | 544 | 84 | ||
XM_015283598.2 | XP_015139084.1 | 4 - 5 | 686 | 84 | ||
XM_015283601.2 | XP_015139087.1 | 4 - 5 | 540 | 84 | ||
XM_015283602.2 | XP_015139088.1 | 4 - 5 | 531 | 84 | ||
XM_015283605.2 | XP_015139091.1 | 4 - 5 | 674 | 84 | ||
XM_015283606.2 | XP_015139092.1 | 4 - 5 | 527 | 84 | ||
XM_015283607.2 | XP_015139093.1 | 4 - 5 | 671 | 84 | ||
XM_025148476.1 | XP_025004244.1 | 4 - 5 | 499 | 84 | ||
XM_025148477.1 | XP_025004245.1 | 4 - 5 | 490 | 84 | ||
XM_025148478.1 | XP_025004246.1 | 4 - 5 | 463 | 84 | ||
XM_025148479.1 | XP_025004247.1 | 4 - 5 | 530 | 84 | ||
GenBank mRNA | AB017437.1 | - | 4 - 5 | 704 | 84 | |
AB035522.1 | - | 4 - 5 | 704 | 84 | ||
AB057641.1 | - | 5 - 6 | 490 | 84 | ||
CR733294.1 | - | 4 - 5 | 621 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
