Assay Details
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205124.1 | NP_990455.1 | 6 - 7 | 939 | 68 | |
GenBank mRNA | D17531.1 | - | 6 - 7 | 939 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
