Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205146.2 | NP_990477.1 | 5 - 6 | 1336 | 77 | |
XM_015278761.1 | XP_015134247.1 | 5 - 6 | 1474 | 77 | ||
GenBank mRNA | X99454.1 | - | 1340 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
