Assay Details
Gene Symbol: PPP4R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006142.1 | NP_001006142.1 | 3 - 4 | 440 | 134 | |
XM_025154495.1 | XP_025010263.1 | 6 - 7 | 6193 | 134 | ||
GenBank mRNA | AJ719288.1 | - | 3 - 4 | 440 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
