Assay Details
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006227.1 | NP_001006227.1 | 9 - 10 | 787 | 81 | |
GenBank mRNA | AJ720776.1 | - | 9 - 10 | 819 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
