Assay Details
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006242.1 | NP_001006242.1 | 3 - 4 | 319 | 88 | |
XM_015285245.2 | XP_015140731.1 | 3 - 4 | 977 | 88 | ||
XM_015285248.2 | XP_015140734.1 | 3 - 4 | 773 | 88 | ||
XM_025148605.1 | XP_025004373.1 | 3 - 4 | 354 | 88 | ||
XM_025148608.1 | XP_025004376.1 | 3 - 4 | 836 | 88 | ||
GenBank mRNA | AJ721012.1 | - | 3 - 4 | 319 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
