Assay Details
Gene Symbol: ADPRHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006312.1 | NP_001006312.1 | 4 - 5 | 723 | 84 | |
XM_025142993.1 | XP_024998761.1 | 4 - 5 | 711 | 84 | ||
GenBank mRNA | AJ720933.1 | - | 4 - 5 | 723 | 84 | |
BX933524.2 | - | 2 - 3 | 217 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
