Assay Details
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006323.1 | NP_001006323.1 | 1299 | 87 | ||
XM_015298930.2 | XP_015154416.1 | 1295 | 87 | |||
XM_025143692.1 | XP_024999460.1 | 2823 | 87 | |||
GenBank mRNA | AJ719588.1 | - | 1299 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
