Assay Details
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006490.2 | NP_001006490.1 | 2 - 3 | 150 | 79 | |
GenBank mRNA | AJ720567.1 | - | 2 - 3 | 151 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
