Assay Details
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006490.2 | NP_001006490.1 | 4 - 5 | 453 | 72 | |
XM_015287491.2 | XP_015142977.1 | 4 - 5 | 480 | 72 | ||
XM_015287492.2 | XP_015142978.1 | 5 - 6 | 525 | 72 | ||
XM_025150798.1 | XP_025006566.1 | 7 - 8 | 633 | 72 | ||
GenBank mRNA | AJ720567.1 | - | 4 - 5 | 454 | 72 | |
CR389417.1 | - | 4 - 5 | 462 | 72 | ||
CR406606.1 | - | 1 - 2 | 241 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
