Assay Details
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006531.1 | NP_001006531.1 | 6 - 7 | 624 | 105 | |
GenBank mRNA | AJ719915.1 | - | 6 - 7 | 624 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
