Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006573.1 | NP_001006573.1 | 13 - 14 | 1520 | 65 | |
XM_015294804.2 | XP_015150290.1 | 12 - 13 | 1421 | 65 | ||
XM_025155117.1 | XP_025010885.1 | 12 - 13 | 1408 | 65 | ||
GenBank mRNA | AJ719414.1 | - | 13 - 14 | 1520 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
