Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006573.1 | NP_001006573.1 | 15 - 16 | 1748 | 63 | |
XM_015294804.2 | XP_015150290.1 | 14 - 15 | 1649 | 63 | ||
XM_025155117.1 | XP_025010885.1 | 14 - 15 | 1636 | 63 | ||
GenBank mRNA | AJ719414.1 | - | 15 - 16 | 1748 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
