Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007083.1 | NP_001007084.1 | 1 - 2 | 128 | 92 | |
GenBank mRNA | AJ621251.1 | - | 1 - 2 | 128 | 92 | |
GU119891.1 | - | 1 - 2 | 128 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
