Assay Details
Gene Symbol: IL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007083.1 | NP_001007084.1 | 2 - 3 | 152 | 64 | |
GenBank mRNA | AJ621251.1 | - | 2 - 3 | 152 | 64 | |
GU119891.1 | - | 2 - 3 | 152 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
