Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204416.1 | NP_989747.1 | 3 - 4 | 239 | 68 | |
GenBank mRNA | M37336.1 | - | 3 - 4 | 239 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
