Assay Details
Gene Symbol: CCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204776.1 | NP_990107.1 | 1 - 2 | 135 | 104 | |
GenBank mRNA | FR874030.1 | - | 1 - 2 | 73 | 104 | |
L34552.1 | - | 1 - 2 | 135 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
