Assay Details
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204778.1 | NP_990109.1 | 14 - 15 | 1779 | 86 | |
XM_015279900.2 | XP_015135386.1 | 15 - 16 | 1959 | 86 | ||
XM_015279904.2 | XP_015135390.1 | 15 - 16 | 1953 | 86 | ||
XM_015279909.2 | XP_015135395.1 | 15 - 16 | 1886 | 86 | ||
GenBank mRNA | AF166094.1 | - | 14 - 15 | 1779 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
